Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, For gas chromatography ECD and FID SupraSolv™ MilliporeSigma™

Catalog No. p-7020418
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle

For gas chromatography ECD and FID SupraSolv™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Color Colorless
Density 1.4g/cm3 at 20°C
Boiling Point 61°C
Quantity 1 L
Odor Sweet
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 211 hPa (20°C)
UN Number 1888
Solubility Information Soluble in water: 8g/L at 20°C
Formula Weight 119.38
Chemical Name or Material Chloroform
Show More Show Less
Hazard Category H302: Harmful if swallowed.
Hazard Statement H315: Causes skin irritation.
Precautionary Statement H319: Causes serious eye irritation.
Recommended Storage 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.