Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform (HPLC), Fisher Chemical™

Catalog No. c607sk1 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 

HPLC mobile phase for analytical and preparative separations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3, 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Absorbance 0.01 AU max., 0.01 AU max. at 280nm, 0.05 AU max. at 270nm, 0.15 AU max. at 254nm, 0.15 AU max. at 260nm, 0.25 AU max. at 255nm, 0.32 AU max. at 250nm, 1.00 AU max. at 245nm, Pass Test
Color Colorless
Density 1.480g/cm³
Boiling Point 61°C
Quantity 1 L
Identification Pass Test
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Refractive Index 1.4430 to 1.4460 (at 25°C)
Residue after Evaporation 2ppm max.
Assay Percent Range 99.8% min by GC
Vapor Pressure 213mbar at 20°C
Viscosity 0.56 mPa/s at 20°C
Water 0.02% max.
Formula Weight 119.38
Percent Purity ≥99.8%
Grade Mobile phase for HPLC applications
Concentration or Composition (by Analyte or Components) Approx. 50ppm Amylene as Preservative
Chemical Name or Material Chloroform, Approx. 50ppm Pentene as Preservative
Show More Show Less
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin. Symptoms of overexposure are dizziness, headache, tiredness, nausea, unconsciousness, cessation of breathing.
<special id
DOTInformation DOT Class 6.1, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.