Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, HPLC Grade, 99.5+% min, stab. with amylene

Catalog No. p-4408234
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES ClC(Cl)Cl

Specifications

Melting Point °C to 63°C
Color Colorless
Density 1.492 g/cm3 at 20°C at 20°C
Boiling Point 60.5°C to 61.5°C
Quantity 1 L
Ignition Point 982°C (1800°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.446
Assay Percent Range >99.5%
Vapor Pressure 158mm Hg at 20°C
Beilstein 1731042
Solubility Information Miscible with diethyl ether,oils,ligroin,alcohol,carbon tetrachloride and carbon disulfide.
Formula Weight 119.38
Grade HPLC
Chemical Name or Material Chloroform
Show More Show Less
Hazard Category H302-H315-H319-H331-H335-H336-H351-H361d-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 200-663-8
RTECSNumber FS9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.