Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform-Isoamyl Alcohol, 24:1, BiotechGrade, Spectrum™ Chemical
SDP

Catalog No. 18601620 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
950 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-601-620 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-620 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C3026950ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C3026

Spectrum™ Chemical Chloroform-Isoamyl Alcohol, 24:1, BiotechGrade - Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Chemical Identifiers

CAS 67-66-3 , 123-51-3 , 64-17-5

Specifications

CAS Max % 90 to 100%, 1 to 5%, 1 to 5%
Quantity 950 mL
Specific Gravity 1.446-1.463
UN Number UN1888
Grade Biotech
Chemical Name or Material Chloroform-Isoamyl Alcohol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.