Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Purified, Reagents

Catalog No. 01337322
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-322 2.5 L
01-337-321 20 L
01-337-320 4 L
01-337-319 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-322 Supplier Reagents Holdings Llc Supplier No. C21755002.5C

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chloroform, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym Trichloromethane, Methyl trichloride
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.5°C
Color Colorless
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.484
Boiling Point 60.5°C to 61.2°C
Grade Pure
Chemical Name or Material Chloroform, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.