Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloroform Solution, NMR Reference Standard, 3% in Acetone-d6 (99.9 atom% D), TMS 0.2%, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
0.235 mL
Specifications
Specifications
Packaging | 3 mm O.D. tube |
CAS | 666-52-4 |
Flash Point | -19°C |
MDL Number | MFCD00000826 |
Type | NMR Reference Standard |
Quantity | 0.235 mL |
Grade | NMR Reference Standard |
Concentration or Composition (by Analyte or Components) | 3% in acetone-d 6 (99.9 atom % D) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction