Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Stabilized, Reagent, ACS, 99.8%, Spectrum™ Chemical
SDP

Catalog No. 18601618
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can

C1220, 67-66-3, CHCl3

Spectrum™ Chemical Chloroform, Stabilized, Reagent, ACS , is a colorless, sweet smelling organic compound comes in the form of a dense liquid and is a trihalomethane. It is also commonly used as laboratory solvent because it is relatively unreactive, conveniently volatile and is miscible with most organic compounds. Although it uses as a reagent are limited, it is used in organic synthesis in order to produce dichlorocarbene by reacting with sodium hydroxide. As an ACS grade Reagent, Spectrum Chemical manufactured chloroform is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

CAS Max % 100%
Quantity 20 L
Packaging Steel Can
Residue after Evaporation 0.001%
UN Number UN1888
Percent Purity 99.8%
Grade ACS
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.