Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloromethane Solution, 200 μg/mL in Methanol, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 mL
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications
Specifications
Packaging | Ampule |
CAS | 74-87-3 |
Flash Point | 9°C |
Linear Formula | CH3 Cl |
MDL Number | MFCD00000872 |
Type | Food and Cosmetic Component Standard |
Quantity | 1 mL |
Formula Weight | 50.49 |
Grade | Analytical Standard |
Concentration or Composition (by Analyte or Components) | 200 μg/mL in methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction