Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethanesulfonyl chloride, 95%

Catalog No. p-7051541
Click to view available options
Quantity:
2 g
10 g
50 g

CAS: 3518-65-8 | CH2Cl2O2S | 148.99 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is synthesis of chloromethanesulfonates and chloromethanesulfonamides. It is also used for conversion of alcohols into the inverted acetates, azides and nitriles, and rearrangement of carbon-carbon and carbon-oxygen bonds under mild conditions, and in electrophilic and radical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3518-65-8
Molecular Formula CH2Cl2O2S
Molecular Weight (g/mol) 148.99
MDL Number MFCD00014727
InChI Key KQDDQXNVESLJNO-UHFFFAOYSA-N
Synonym methanesulfonyl chloride, chloro, chloromethylsulfonyl chloride, chloromethanesulphonyl chloride, chlormethansulfochlorid, chlormethansulfonylchlorid, chloromesyl chloride, chloromethyl sulfonyl chloride, chlormethansulfochlorid czech, chlormethansulfonylchlorid czech, chlorid kyseliny chlormethansulfonove
PubChem CID 77054
IUPAC Name chloromethanesulfonyl chloride
SMILES ClCS(Cl)(=O)=O

Specifications

Color Colorless to Yellow
Density 1.67 g/mL
Boiling Point 78°C to 81°C (20 mmHg)
Odor Acidic
Assay Percent Range 98%
Linear Formula ClCH2SO2Cl
Quantity 2 g
UN Number UN3265
Beilstein 1811761
Sensitivity Moisture sensitive
Solubility Information It is soluble in dichloromethane,pyridine,and most organic solvents. It is insoluble in water.
Formula Weight 149
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloromethanesulfonyl chloride
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 222-526-1
RTECSNumber PB2800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.