Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl Chlorosulfonate 98.0+%, TCI America™
SDP

Catalog No. C266025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C266025G 25 g
C26605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C266025G Supplier TCI America Supplier No. C266025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49715-04-0
Molecular Formula CH2Cl2O3S
Molecular Weight (g/mol) 164.984
MDL Number MFCD00041547
InChI Key PJBIHXWYDMFGCV-UHFFFAOYSA-N
Synonym chloromethyl chlorosulfate, chloromethyl chlorosulphate, chloromethylchlorosulfate, chloromethyl chlorosulfonate, chloromethyl chloranesulfonate, unii-n615783bnf, chloromethyl sulfochloridate, chlorosulfuric acid, chloromethyl ester, chloro chlorosulfonyloxy methane, chlorosulfuric acid chloromethyl ester
PubChem CID 2733556
IUPAC Name chloro(chlorosulfonyloxy)methane
SMILES C(OS(=O)(=O)Cl)Cl

Specifications

Color Yellow
Boiling Point 50°C
Quantity 25 g
UN Number 3265
Formula Weight 164.98
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Chloromethyl Chlorosulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.