Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chloromethyl)dimethylchlorosilane, 98%

Catalog No. p-7022226
Click to view available options
Quantity:
10 g
25 g
100 g

CAS: 1719-57-9 | C3H8Cl2Si | 143.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-57-9
Molecular Formula C3H8Cl2Si
Molecular Weight (g/mol) 143.08
MDL Number MFCD00000875
InChI Key ITKVLPYNJQOCPW-UHFFFAOYSA-N
Synonym chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane
PubChem CID 74393
IUPAC Name chloro-(chloromethyl)-dimethylsilane
SMILES C[Si](C)(Cl)CCl

Specifications

Boiling Point 110°C to 113°C
Quantity 10 g
Linear Formula ClCH2Si(CH3)2Cl
UN Number UN2985
Beilstein 1731933
Flash Point 21°C (69°F)
Formula Weight 143.09
Percent Purity 98%
Odor Acrid
Refractive Index 1.437
Sensitivity Moisture sensitive
Density 1.082
Chemical Name or Material (Chloromethyl)dimethylchlorosilane
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 217-006-6
RTECSNumber VV2095000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.