Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl Ethyl Ether 96.0+%, TCI America™
SDP

Catalog No. C0201100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0201100G 100 g
C020125G 25 g
C0201500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0201100G Supplier TCI America Supplier No. C0201100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3188-13-4
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.54
MDL Number MFCD00000887
InChI Key FCYRSDMGOLYDHL-UHFFFAOYSA-N
Synonym chloromethyl ethyl ether, chloromethoxy ethane, ethane, chloromethoxy, chloromethylethylether, ethoxymethyl chloride, ethyl chloromethyl ether, ethoxychloromethane, ether, chloromethyl ethyl, ethoxy methyl chloride
PubChem CID 18523
IUPAC Name (chloromethoxy)ethane
SMILES CCOCCl

Specifications

Color Yellow
Boiling Point 80°C
Quantity 100 g
UN Number 2354
Formula Weight 94.54
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl Ethyl Ether
EINECSNumber (2)-3725
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.