Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl methyl sulfide, 94%

Catalog No. AAL1085818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1085818 50 g
AAL1085809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1085818 Supplier Thermo Scientific Chemicals Supplier No. L1085818
Only null left
Add to Cart

CAS: 2373-51-5 | C2H5ClS | 96.57 g/mol

Chloromethyl methyl sulfide is used as a methylene transfer reagent for iron(II) mediated cyclopropanation and for carbonyl and aromatic compounds. It protects alcohols as their methylthiomethyl (MTM) ethers by reaction, e.g. with NaH/NaI in DME. It is also used for the preparation of MTM esters of carboxylic acids, e.g. by treatment of the K salt of the acid in the presence of NaI and 18-crown-6 and for reaction with cyclopentadienyl iron dicarbonyl dimer in the formation of an iron-containing methylene transfer agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloromethyl methyl sulfide is used as a methylene transfer reagent for iron(II) mediated cyclopropanation and for carbonyl and aromatic compounds. It protects alcohols as their methylthiomethyl (MTM) ethers by reaction, e.g. with NaH/NaI in DME. It is also used for the preparation of MTM esters of carboxylic acids, e.g. by treatment of the K salt of the acid in the presence of NaI and 18-crown-6 and for reaction with cyclopentadienyl iron dicarbonyl dimer in the formation of an iron-containing methylene transfer agent.

Solubility
Soluble in acetonitrile, ether and dichloromethane. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2373-51-5
Molecular Formula C2H5ClS
Molecular Weight (g/mol) 96.57
MDL Number MFCD00000923
InChI Key JWMLCCRPDOIBAV-UHFFFAOYSA-N
Synonym chloromethyl methyl sulfide, chlorodimethyl sulfide, methylthiomethyl chloride, chloro methylthio methane, methane, chloro methylthio, chlorodimethyl thioether, chloromethyl methyl thioether, monochlorodimethyl sulfide, chloro methylsulfanyl methane, clorodimetilsolfuro
PubChem CID 16916
IUPAC Name chloro(methylsulfanyl)methane
SMILES CSCCl

Specifications

Density 1.16
Boiling Point 104°C to 105°C
Flash Point 17°C (62°F)
Odor Stench
Refractive Index 1.498
Quantity 50 g
UN Number UN1993
Beilstein 1730851
Sensitivity Moisture sensitive
Solubility Information Soluble in acetonitrile,ether and dichloromethane. Insoluble in water.
Formula Weight 96.58
Percent Purity 95%
Chemical Name or Material Chloromethyl methyl sulfide
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 219-148-4
RTECSNumber WQ3700000
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.