Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethylated Polystyrene: 1% Divinylbenzene Copolymer Beads, Thermo Scientific™

Catalog No. AC404740100
Click to view available options
Quantity:
10 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70024-51-0
Molecular Formula C12H23NO4
Molecular Weight (g/mol) 245.319
MDL Number MFCD00146406
InChI Key FCGHBWUKOGKLKO-UNXYVOJBSA-N
PubChem CID 122172946
IUPAC Name 2-[(3S)-heptan-3-yl]-5-methyl-5-nitro-1,3-dioxane
SMILES CCCCC(CC)C1OCC(CO1)(C)[N+](=O)[O-]

Specifications

Color White
Infrared Spectrum Authentic
Mesh Size 100 to 200 mesh
Quantity 10 g
Physical Form Powder
Chemical Name or Material Chloromethylated polystyrene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.