Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chloromethylene)dimethylammonium chloride, 96%

Catalog No. p-88336
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 3724-43-4 | C3H7Cl2N | 128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3724-43-4
Molecular Formula C3H7Cl2N
Molecular Weight (g/mol) 128
MDL Number MFCD00011868
InChI Key QQVDYSUDFZZPSU-UHFFFAOYSA-M
Synonym arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride
PubChem CID 77311
IUPAC Name chloromethylidene(dimethyl)azanium;chloride
SMILES C[N+](=CCl)C.[Cl-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 139°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Argentometry)
Packaging Glass bottle
Linear Formula ClCH=N(CH3)2Cl
Quantity 5 g
Fieser 04,186; 05,251; 06,220; 07,422; 08,186; 09,514; 10,457; 12,564; 13,341
Solubility Information Solubility in water: reacts. Other solubilities: 28g/100mL chloroform,slightly soluble in other solvents
Formula Weight 128
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material (Chloromethylene)dimethylammonium chloride, 96%
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May damage the unborn child.
Reacts violently with water.
missing translation for 'healthHazard3' GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.