Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorophenol Red Indicator, 0.04%, Reagents

Catalog No. 01335691
Click to view available options
Quantity:
120 mL
250 mL

Catalog No. 01-335-691

Supplier: Reagents Holdings Llc CC011000120A

Only null left
Add to Cart
Add to Cart

Chlorophenol Red Indicator, 0.04%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Chlorophenol Red Indicator, 0.04%
CAS 7732-18-5, 123333-64-2
Boiling Point 100°C
Percent Purity 0.04%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0001
Color Red
Quantity 120 mL
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.