Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloroplatinate Solution, 3000 Color Units, Reagents
Click to view available options
Quantity:
1 L
Specifications
Specifications
Chemical Name or Material | Chloroplatinate Solution, 3000 Color Units |
Packaging | Glass Bottle |
CAS | 7732-18-5, 7647-01-0, 16921-30-5 |
Specific Gravity | 1.027 |
Physical Form | Liquid |
Color | Orange |
Solubility | Completely miscible |
Quantity | 1 L |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction