Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroplatinic Acid Hexahydrate, ACS Reagent, ≥37.50% Pt basis, Honeywell™

Catalog No. 60045126 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle

ACS Reagent,≥37.50% Pt basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Physical Form Powder
Quantity 25 g
Linear Formula H2PtCl6 · 6H2O
UN Number UN2507
Formula Weight 517.9g/mol
Percent Purity ≥37.50% (Pt basis)
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Chloroplatinic Acid Hexahydrate
Hazard Statement P261-P280-P301 + P310-P305 + P351 + P338-P310
Precautionary Statement H301-H314-H317-H334
DOTInformation 8/PGIII
EINECSNumber 241-010-7
RTECSNumber TP1510000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.