Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorosulfonic Acid

Catalog No. 571024100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
5 g
100 g
1 kg
4 x 100 g
Danger

Catalog No. 57-102-4100G

Supplier: Sigma Aldrich 571024100G

Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorosulfonic acid is a strong acid that is synthesized on an industrial scale by reacting sulfur trioxide and dry hydrogen chloride gas in an equimolar ratio. It can act as a sulfonating, dehydrating, oxidizing and chlorinating agent. It is reported to be a better electrophilic olefin cyclization agent in comparison to other sulfonic acids. Its infrared spectra in the solid, liquid and gaseous state have been recorded in the region 4000-400cm-1.Chlorosulfonic acid may be used in the following processes: Synthesis of di-n-butyl ammonium chlorosulfonate, a secondary amine ionic liquid.To prepare sulfonated polyethersulfone (SPES) with enhanced hydrophilic property.To attach sulfate group to silk fibroin for improving its anticoagulant ability.As a catalyst to prepare substituted coumarin, via von Pechmann condensation between phenol and +a-ketoester.As a substitute to acid catalyst in Biginelli reaction to prepare 3,4dihydropyrimidines.

Specifications

CAS 7790-94-5
Density 1.753 g/mL at 25°C
Boiling Point 151°C to 152°C (755 mmHg, lit.)
Molecular Formula HClO3S
Refractive Index n20/D 1.433
Linear Formula ClSO3H
MDL Number MFCD00011523
Quantity 100 g
Molecular Weight (g/mol) 116.52 g/mol
Biological Activity Respiratory System
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.