Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorothiazide 97.0+%, TCI America™
SDP

Catalog No. C225925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C225925G 25 g
C22595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C225925G Supplier TCI America Supplier No. C225925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58-94-6
Molecular Formula C7H6ClN3O4S2
Molecular Weight (g/mol) 295.71
MDL Number MFCD00058576
InChI Key JBMKAUGHUNFTOL-UHFFFAOYSA-N
Synonym chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit
PubChem CID 2720
ChEBI CHEBI:3640
IUPAC Name 6-chloro-1,1-dioxo-4H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl

Specifications

Melting Point 348°C
Color White-Yellow
Quantity 25 g
Formula Weight 295.71
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorothiazide
RTECSNumber DK9450000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.