Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorotriethylsilane, 1M solution in THF, AcroSeal™

Catalog No. AC433251000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 994-30-9 | C6H15ClSi | 150.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 994-30-9, 109-99-9
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00000507
InChI Key DCFKHNIGBAHNSS-UHFFFAOYSA-N
Synonym triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
PubChem CID 13819
IUPAC Name chloro(triethyl)silane
SMILES CC[Si](Cl)(CC)CC

Specifications

Color Colorless
Density 0.8860g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (C2H5)3SiCl
Quantity 100 mL
Specific Gravity 0.886
Merck Index 15,2179
Formula Weight 150.72
Physical Form Liquid
Chemical Name or Material Chlorotriethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
Contact with water liberates toxic gas.
May form explosive per
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 213-615-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.