Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorotriphenylmethane, 98%

Catalog No. aaa1179914
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g

CAS: 76-83-5 | C19H15Cl | 278.78 g/mol

Chlorotriphenylmethane is used as laboratory chemical. It acts as a protecting reagent for thiols, alcohols and amines. It is used in organic synthesis for initiating trityl group protection and for preparing S-trityl-L-cysteine (STLC), a inhibitor of Eg5 prevents mitotic progression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriphenylmethane is used as laboratory chemical. It acts as a protecting reagent for thiols, alcohols and amines. It is used in organic synthesis for initiating trityl group protection and for preparing S-trityl-L-cysteine (STLC), a inhibitor of Eg5 prevents mitotic progression.

Solubility
Insoluble in water. Soluble in chloroform, hexane, benzene, ether, acetone and tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-83-5
Molecular Formula C19H15Cl
Molecular Weight (g/mol) 278.78
MDL Number MFCD00000813,MFCD00284810
InChI Key JBWKIWSBJXDJDT-UHFFFAOYSA-N
Synonym trityl chloride, triphenylmethyl chloride, triphenylchloromethane, chlorotriphenylmethane, chloromethanetriyl tribenzene, triphenylmethylchloride, methane, chlorotriphenyl, triphenyl chloromethane, chlorodiphenylmethyl benzene, benzene, 1,1',1-chloromethylidyne tris
PubChem CID 6456
IUPAC Name [chloro(diphenyl)methyl]benzene
SMILES ClC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 110°C to 114°C
Boiling Point 230°C to 235°C (20 mmHg)
Linear Formula (C6H5)3CCl
Quantity 25 g
UN Number UN3261
Beilstein 397363
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in chloroform,hexane,benzene,ether,acetone and tetrahydrofuran.
Formula Weight 278.78
Percent Purity 98%
Chemical Name or Material Chlorotriphenylmethane
Show More Show Less
missing translation for 'healthHazard1' H314-H335
missing translation for 'healthHazard2' GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
missing translation for 'healthHazard3' P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 200-986-4
RTECSNumber PA6450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.