Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroxylenol, USP, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18601635
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle

CH136, 88-04-0, C8H9ClO

Spectrum™ Chemical Chloroxylenol, USP is a broad spectrum antimicrobial chemical compound used to control bacteria, algae, fungi and virus. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 88-04-0
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
InChI Key OSDLLIBGSJNGJE-UHFFFAOYSA-N
IUPAC Name 4-chloro-3,5-dimethylphenol
SMILES CC1=CC(O)=CC(C)=C1Cl

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 50 kg
Percent Purity 98.5%
Grade USP
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.