Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorpromazine hydrochloride, 100.9%, MP Biomedicals™

Catalog No. ICN19032625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19032625 25 g
ICN19032605 5 g
ICN19032680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19032625 Supplier MP Biomedicals, Inc Supplier No. 0219032625
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69-09-0
Molecular Formula C17H20Cl2N2S
Molecular Weight (g/mol) 355.321
InChI Key FBSMERQALIEGJT-UHFFFAOYSA-N
Synonym 2-Chloro-10-[3-dimethylamino-propyl] phenothiazine, 2-Chloro-N, N- dimethyl-10H- phenothiazine-10- propanamine hydrochloride
PubChem CID 6240
ChEBI CHEBI:3649
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2SC3=C1C=C(C=C3)Cl.Cl

Specifications

Assay Percent Range 1.009
Physical Form Powder/Solution
Color Colorless or White
Quantity 25 g
Formula Weight 355.3
Chemical Name or Material Chlorpromazine hydrochloride
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.