Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlorpyrifos Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP288858
Description
Chlorpyrifos Polyclonal specifically detects Chlorpyrifos in Non-species specific samples. It is validated for ELISA, Immunoaffinity Purification.Specifications
Chlorpyrifos | |
Polyclonal | |
ELISA 1:500, Immunoaffinity Purification | |
chlorpyrifos-ethyl, Clorpyrifos | |
BSA-Chlorpyrifos conjugate | |
1 mL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA, Affinity Purification | |
Unconjugated | |
PBS | |
Rabbit | |
Caprylic acid precipitation | |
RUO | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction