Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chlorpyrifos Monoclonal Antibody (3E4)
GREENER_CHOICE

Catalog No. PIMA549719
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549719 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549719 Supplier Invitrogen™ Supplier No. MA549719
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Chlorpyrifos is an insecticide belonging to the group of organophosphothioates. It is used in the culture of vegetables and in citrus orchards. Chlorpyrifos is an acetylcholinesterase inhibitor and as such having a highly toxic effect on the nervous system. There is evidence that chlorpyrifos has an effect on the foetus, which may lead to behavioural disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlorpyrifos
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3E4
Concentration 2.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of CPF hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.