Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur.

Catalog No. p-4229149
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family. | CAS: 64-72-2 | C22H23ClN2O8·HCl | 515.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General features
• Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
• Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
• A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
• Can work as an antiprotozoal and antifungal


Applications
• Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
• Intended for research use only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22H23ClN2O8·HCl
Molecular Weight (g/mol) 515.33
MDL Number MFCD00082440
InChI Key QYAPHLRPFNSDNH-CIVPRPTRSA-N
Synonym chlortetracycline hydrochloride, 4-epi-chlortetracycline hydrochloride, 2-amino hydroxy methylidene-7-chloro-4-dimethylamino-6,10,11,12a-tetrahydroxy-6-methyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride, 2-azanyl oxidanyl methylidene-7-chloranyl-4-dimethylamino-6-methyl-6,10,11,12a-tetrakis oxidanyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride
PubChem CID 66577600
IUPAC Name (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Absorbance 0.40 max. (460nm)
Melting Point 220.0°C
Color Yellow
pH 2.3 to 3.3 (1% H2O, CO2 free)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Merck Index 15,2198
Specific Rotation Condition − 242.50 (20.00°C c=0.5,water)
Specific Rotation − 242.50
Solubility Information Solubility in water: 8.6g/L (28°C). Other solubilities: practically insoluble in acetone,ether,,chloroform,dioxane and propylene glycol,17.4g/L methanol (28°C),1.7g/L ethanol (28°C),soluble in solutions of alkali hydroxides and,carbonat
Formula Weight 515.33
Percent Purity 94.5 to 102.0% (sum of the contents of chlortetra cycline hydrochloride and tetracycline hydrochloride, dry substance), ≥89.5% (chlortetracycline hydrochloride, dry substance), 8% max. (tetracycline hydrochloride, dry substance)
Physical Form Powder
Chemical Name or Material Chlortetracycline hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 200-591-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.