Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHML Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556213 Supplier Invitrogen™ Supplier No. PA556213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YASQDMEDNV EEIGALQKNP SLGVSNTFTE VLDSALPEES QLSYFNSDEM PAKHTQKSDT EISLE Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 39%.

Binds unprenylated Rab proteins, presents it to the catalytic component B, and remains bound to it after the geranylgeranyl transfer reaction. The component A may be regenerated by transferring its prenylated Rab to a protein acceptor. Less effective than REP-1 in supporting prenylation of Rab3 family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHML
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHML
Gene Accession No. P26374
Gene Alias AI450814; CHM like, Rab escort protein 2; Chml; choroideraemia-like protein; choroideremia-like; choroideremia-like (Rab escort protein 2); choroideremia-like protein; E030003F13Rik; rab escort protein 2; rab escort protein-2; rab proteins geranylgeranyltransferase component A 2; REP2; REP-2
Gene Symbols CHML
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHML. Recombinant protein control fragment (Product #RP-94561).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1122
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.