Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544773 Supplier Invitrogen™ Supplier No. PA544773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KIKKAIQKGN MEVARIHAEN AIRQKNQAVN FLRMSARVDA VAARVQTAVT Sequence homology: Cow: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

CHMP1B belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHMP1B
Gene Accession No. Q7LBR1
Gene Alias 2810405I11Rik; C10orf2; C18orf2; C18-ORF2; Charged multivesicular body protein 1b; charged multivesicular body protein 1b-1; CHMP1.5; CHMP1.5 protein; chmp1b; Chmp1b1; CHMP1b-1; chromatin modifying protein 1B; chromatin-modifying protein 1b; Chromatin-modifying protein 1b-1; fb10c03; hVps46-2; hypothetical protein LOC336426; Similar to RIKEN cDNA 2810405I11 gene; vacuolar protein sorting 46-2; vacuolar protein sorting-associated protein 46-2; Vps46-2; Vps46B; wu:fb10c03; zgc:56134
Gene Symbols CHMP1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CHMP1B (aa 35-84).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57132
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.