Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHMP1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563997
Description
Immunogen sequence: QNQVDMLLQE MADEAGLDLN MELPQGQTGS VGTSVASAEQ DELSQRLARL RDQV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 96%.
CHMP1B belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 ).
Specifications
CHMP1B | |
Polyclonal | |
Unconjugated | |
CHMP1B | |
2810405I11Rik; C10orf2; C18orf2; C18-ORF2; Charged multivesicular body protein 1b; charged multivesicular body protein 1b-1; CHMP1.5; CHMP1.5 protein; chmp1b; Chmp1b1; CHMP1b-1; chromatin modifying protein 1B; chromatin-modifying protein 1b; Chromatin-modifying protein 1b-1; fb10c03; hVps46-2; hypothetical protein LOC336426; Similar to RIKEN cDNA 2810405I11 gene; vacuolar protein sorting 46-2; vacuolar protein sorting-associated protein 46-2; Vps46-2; Vps46B; wu:fb10c03; zgc:56134 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7LBR1 | |
CHMP1B | |
Recombinant protein corresponding to Human CHMP1B. Recombinant protein control fragment (Product #RP-103426). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction