Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP1B Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1974MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1974MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1974MI Supplier MilliporeSigma Supplier No. ABE1974
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-CHMP1B is a highly specific rabbit polyclonal antibody that targets charged multivesicular body protein 1b and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP1B
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CHMP1B,C18orf2
Host Species Rabbit
Immunogen GST-tagged full length recombinant human Charged multivesicular body protein 1b.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_065145
Test Specificity This polyclonal antibody detects Charged multivesicular body protein 1b in human cells. Human CHMP1B displays 99% and 98% homology with bovine and mouse CHMP1B, respectively.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.