Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP2A Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297282MFV610
Description
CHMP2A Polyclonal antibody specifically detects CHMP2A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CHMP2A | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
BC-2, BC2VPS2 homolog A, CHMP2a, CHMP2hVps2-1, chromatin modifying protein 2A, Chromatin-modifying protein 2a, Putative breast adenocarcinoma marker BC-2, Vacuolar protein sorting-associated protein 2-1, vps2-1, VPS2Aputative breast adenocarcinoma marker (32kD), VPS2charged multivesicular body protein 2a | |
Produced in rabbits immunized with E. coli-derived Human CHMP2A fragment. | |
0.1 mL | |
Autophagy, Cell Biology, Signal Transduction | |
27243 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction