Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHMP2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559831
Description
Immunogen sequence: MDLLFGRRKT PEELLRQNQR ALNRAMRELD RERQKLETQE KKIIADIKKM AKQGQMDAVR IMAKDLVRTR RYVRKFVLMR A Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
CHMP2A belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A is required for both MVB formation and regulation of cell cycle progression.
Specifications
CHMP2A | |
Polyclonal | |
Unconjugated | |
CHMP2A | |
1500016L11Rik; BC2; BC-2; Charged multivesicular body protein 2a; CHMP2; CHMP2A; chromatin modifying protein 2A; chromatin-modifying protein 2a; fj10d05; hypothetical protein LOC523847; mVps2; putative breast adenocarcinoma marker; putative breast adenocarcinoma marker (32kD); putative breast adenocarcinoma marker BC-2; RGD1305050; Vacuolar protein sorting-associated protein 2; vacuolar protein sorting-associated protein 2-1; vacuolar protein-sorting-associated protein 2; VPS2; VPS2 homolog A; vps2-1; VPS2A; wu:fi18h03; wu:fj10d05; zgc:55361 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27243 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43633 | |
CHMP2A | |
Recombinant protein corresponding to Human CHMP2A. Recombinant protein control fragment (Product #RP-99060). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction