Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587855 Supplier Invitrogen™ Supplier No. PA587855
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDLLFGRRKT PEELLRQNQR ALNRAMRELD RERQKLETQE KKIIADIKKM AKQGQMDAVR IMAKDLVRTR RYVRKFVLMR ANIQAVSLKI QTLKSNNSMA QAMKGVTKAM GTMNRQLKLP QIQKIMMEFE RQAEIMDMKE EMMNDAIDDA MGDEEDEEES DAVVSQVLDE LGLSLTDELS NLPSTGGSLS VAAGGKKAEA AASALADADA DLEERLKNLR RD; Positive Samples: A-431, HepG2, SW480; Cellular Location: Cytoplasmic side, Late endosome membrane, Peripheral membrane protein.

CHMP2A belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A is required for both MVB formation and regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP2A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHMP2A
Gene Accession No. O43633
Gene Alias 1500016L11Rik; BC2; BC-2; Charged multivesicular body protein 2a; CHMP2; CHMP2A; chromatin modifying protein 2A; chromatin-modifying protein 2a; fj10d05; hypothetical protein LOC523847; mVps2; putative breast adenocarcinoma marker; putative breast adenocarcinoma marker (32kD); putative breast adenocarcinoma marker BC-2; RGD1305050; Vacuolar protein sorting-associated protein 2; vacuolar protein sorting-associated protein 2-1; vacuolar protein-sorting-associated protein 2; VPS2; VPS2 homolog A; vps2-1; VPS2A; wu:fi18h03; wu:fj10d05; zgc:55361
Gene Symbols CHMP2A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-222 of human CHMP2A (NP_0552681).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27243
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.