Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP256956
Description
CHMP2B Polyclonal specifically detects CHMP2B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CHMP2B | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
charged multivesicular body protein 2b, CHMP2.5VPS2BVPS2-2, CHMP2b, chromatin modifying protein 2B, Chromatin-modifying protein 2b, DKFZP564O123, DMT1, hVps2-2, Vacuolar protein sorting-associated protein 2-2, vacuolar protein-sorting-associated protein 2-2, VPS2 homolog B, Vps2-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
25978 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
CHMP2B | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TVDDVIKEQNRELRGTQRAIIRDRAALEKQE | |
100 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction