Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP2B Antibody (SR1463), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32234450UL
Description
CHMP2B Monoclonal antibody specifically detects CHMP2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
CHMP2B | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR1463 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
charged multivesicular body protein 2b, CHMP2.5VPS2BVPS2-2, CHMP2b, chromatin modifying protein 2B, Chromatin-modifying protein 2b, DKFZP564O123, DMT1, hVps2-2, Vacuolar protein sorting-associated protein 2-2, vacuolar protein-sorting-associated protein 2-2, VPS2 homolog B, Vps2-2 | |
A synthesized peptide derived from human CHMP2B (Uniprot #: Q9UQN3) | |
50 μg | |
Autophagy, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction | |
25978 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction