Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531128 Supplier Invitrogen™ Supplier No. PA531128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (100%), Xenopus laevis (93%), Pig (99%), Chicken (95%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CHMP2B, also known as CHMP2.5 or vacuolar protein-sorting-associated protein 2-2, is a 213 amino acid cytosolic protein. Widely expressed in brain, heart, skeletal muscle, small intestine, pancreas, lung, placenta and leukocytes, CHMP2B associates directly with CHMP2A and vps4 for the disassembly of the ESCRT-III complex. Defects in the gene encoding CHMP2B have been shown to cause chromosome 3-linked frontotemporal dementia (FTD3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP2B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Chmp2b
Gene Accession No. Q9UQN3
Gene Alias 1190006E07Rik; ALS17; CGI-84; Charged multivesicular body protein 2b; charged multivesicular body protein 2Bb; CHMP family; CHMP2.5; CHMP2B; chmp2bb; chmp2bl; chromatin modifying protein 2B; chromatin modifying protein 2B, like; chromatin modifying protein 2Bb; chromatin-modifying protein 2b; DMT1; hVps2-2; hypothetical protein LOC615954; hypothetical protein zgc:77025; RGD1306781; si:ch211-106g5.3; Vacuolar protein sorting-associated protein 2-2; vacuolar protein-sorting-associated protein 2-2; VPS2 homolog B; VPS2-2; VPS2B; zgc:77025
Gene Symbols Chmp2b
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CHMP2B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.