Learn More
Invitrogen™ CHMP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587794
Description
Immunogen sequence: MGLFGKTQEK PPKELVNEWS LKIRKEMRVV DRQIRDIQRE EEKVKRSVKD AAKKGQKDVC IVLAKEMIRS RKAVSKLYAS KAHMNSVLMG MKNQLAVLRV AGSLQKSTEV MKAMQSLVKI PEIQATMREL SKEMMKAGII EEMLEDTFES MDDQEEMEEE AEMEIDRILF EITAGALGKA PSKVTDALPE PEPPGAMAAS EDEEEEEEAL EAMQSRLATL RS; Positive Samples: BT-474, LO2, 293T, SW480; Cellular Location: Cytoplasm, Endosome, Late endosome membrane, Lipid-anchor, Membrane, cytosol.
Probable core component of the endosomal sorting required for transport complex III (ESCRT-III) which is involved in multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. MVBs contain intraluminal vesicles (ILVs) that are generated by invagination and scission from the limiting membrane of the endosome and mostly are delivered to lysosomes enabling degradation of membrane proteins, such as stimulated growth factor receptors, lysosomal enzymes and lipids. The MVB pathway appears to require the sequential function of ESCRT-O, -I,-II and -III complexes. ESCRT-III proteins mostly dissociate from the invaginating membrane before the ILV is released. The ESCRT machinery also functions in topologically equivalent membrane fission events, such as the terminal stages of cytokinesis and the budding of enveloped viruses (HIV-1 and other lentiviruses). ESCRT-III proteins are believed to mediate the necessary vesicle extrusion and/or membrane fission activities, possibly in conjunction with the AAA ATPase VPS4. Selectively binds to phosphatidylinositol 3,5-bisphosphate PtdIns(3,5)P2 and PtdIns(3,4)P2 in preference to other phosphoinositides tested. Involved in late stages of cytokinesis. Plays a role in endosomal sorting/trafficking of EGF receptor. Isoform 2 prevents stress- mediated cell death and accumulation of reactive oxygen species when expressed in yeast cells.
Specifications
CHMP3 | |
Polyclonal | |
Unconjugated | |
CHMP3 | |
25.1; 25.1 protein; 4921505F14Rik; 9130011K15Rik; CGI149; CGI-149; CGI-49; Charged multivesicular body protein 3; CHMP family, member 3; CHMP3; Chromatin-modifying protein 3; D6Ertd286e; hVps24; NEDF; Neuroendocrine differentiation factor; rVps24p; vacuolar protein sorting 24; vacuolar protein sorting 24 homolog; vacuolar protein sorting-associated protein 24; vacuolar protein-sorting-associated protein 24; Vps24; Vps24p protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51652, 66700 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CQ10, Q9Y3E7 | |
CHMP3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-222 of human VPS24 (NP_0571631). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.