Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP3 Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS166825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1668-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1668-25UL Supplier MilliporeSigma Supplier No. ABS166825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-CHMP3, Cat. No. ABS1668, is a rabbit polyclonal antibody that detects Charged multivesicular body protein 3 and has been tested for use in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP3
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Charged multivesicular body protein 3;Chromatin-modifying protein 3;Neuroendocrine differentiation factor;Vacuolar protein sorting-associated protein 24;hVps24
Gene Symbols CHMP3;CGI149;NEDF;VPS24;CGI-149
Host Species Rabbit
Immunogen A full-length human recombinant charged multivesicular body protein 3 (CHMP3).
Molecular Weight of Antigen ∽32 kDa observed; 25.07 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Charged multivesicular body protein 3 in human cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Product Type Antibodies
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.