Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP4A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | CHMP4A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17992020
![]() |
Novus Biologicals
NBP17992020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179920
![]() |
Novus Biologicals
NBP179920 |
100 μL |
Each for $487.50
|
|
|||||
Description
CHMP4A Polyclonal specifically detects CHMP4A in Human samples. It is validated for Western Blot.Specifications
CHMP4A | |
Polyclonal | |
Rabbit | |
NP_054888 | |
29082 | |
The immunogen for this antibody is CHMP4A. Peptide sequence ELEQEELAQELLNVGDKEEEPSVKLPSVPSTHLPAGPAPKVDEDEEALKQ. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C14orf123, charged multivesicular body protein 4a, CHMP4, CHMP4a, CHMP4B, chromatin modifying protein 4A, Chromatin-modifying protein 4a, chromosome 14 open reading frame 123, FLJ61658, hSnf-1, HSPC134, hVps32-1, MGC142093, MGC142095, SHAX2, SNF7, SNF7 homolog associated with Alix-2, Snf7 homologue associated with Alix 2, SNF7-1, Vacuolar protein sorting-associated protein 32-1, VPS32-1, VPS32A | |
CHMP4A | |
IgG | |
29 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title