Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP4A Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549410

Catalog No. PA549410


Only null left
Add to Cart

Description

Description

CHMP4A Polyclonal Antibody for Western Blot

Probable core component of the endosomal sorting required for transport complex III (ESCRT-III) which is involved in multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. MVBs contain intraluminal vesicles (ILVs) that are generated by invagination and scission from the limiting membrane of the endosome and mostly are delivered to lysosomes enabling degradation of membrane proteins, such as stimulated growth factor receptors, lysosomal enzymes and lipids. The MVB pathway appears to require the sequential function of ESCRT-O, -I,-II and -III complexes. ESCRT-III proteins mostly dissociate from the invaginating membrane before the ILV is released. The ESCRT machinery also functions in topologically equivalent membrane fission events, such as the terminal stages of cytokinesis and the budding of enveloped viruses (HIV-1 and other lentiviruses). ESCRT-III proteins are believed to mediate the necessary vesicle extrusion and/or membrane fission activities, possibly in conjunction with the AAA ATPase VPS4. When overexpressed, membrane-assembled circular arrays of CHMP4A filaments can promote or stabilize negative curvature and outward budding. Via its interaction with PDCD6IP involved in HIV-1 p6- and p9-dependent virus release.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHMP4A
Polyclonal
Unconjugated
CHMP4A
C14orf123; CDA04; Charged multivesicular body protein 4a; CHMP4; CHMP4A; CHMP4B; chromatin modifying protein 4A; Chromatin-modifying protein 4a; hSnf-1; HSPC134; hVps32-1; SHAX2; SNF7; SNF7 homolog associated with Alix-2; Snf7 homologue associated with Alix 2; SNF7-1; Vacuolar protein sorting-associated protein 32-1; VPS32-1; VPS32A
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
29082
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9BY43
CHMP4A
KLH conjugated synthetic peptide between 205-239 amino acids from the C-terminal region of human CHMP4A.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.