Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP4A Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306005IR
Description
CHMP4A Polyclonal specifically detects CHMP4A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CHMP4A | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C14orf123, charged multivesicular body protein 4a, CHMP4, CHMP4a, CHMP4B, chromatin modifying protein 4A, Chromatin-modifying protein 4a, chromosome 14 open reading frame 123, FLJ61658, hSnf-1, HSPC134, hVps32-1, MGC142093, MGC142095, SHAX2, SNF7, SNF7 homolog associated with Alix-2, Snf7 homologue associated with Alix 2, SNF7-1, Vacuolar protein sorting-associated protein 32-1, VPS32-1, VPS32A | |
Produced in rabbits immunized with E. coli-derived Human CHMP4A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
29082 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction