Learn More
Invitrogen™ CHMP4B Polyclonal Antibody

Description
This antibody is predicted to react with chicken, mouse, Xenopus and zebrafish based on sequence homology.
Specifications
Specifications
| Antigen | CHMP4B |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CHMP4B |
| Gene Accession No. | Q9H444 |
| Gene Alias | 2010012F05Rik; C20orf178; C76846; charged multivesicular body protein 4b; CHMP4A; CHMP4B; chromatin modifying protein 4B; chromatin-modifying protein 4b; CTPP3; CTRCT31; dJ553F4.4; hSnf7-2; hVps32-2; RGD1309846; RGD1565889; Shax1; SNF7; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; Snf7-2; Vacuolar protein sorting-associated protein 32-2; vacuolar protein-sorting-associated protein 32-2; Vps32-2; VPS32B |
| Gene Symbols | CHMP4B |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.