Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559552 Supplier Invitrogen™ Supplier No. PA559552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVFGKLFGAG GGKAGKGGPT PQEAIQRLRD TEEMLSKK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

CHMP4B is a member of the chromatin-modifying protein/charged multivesicular body protein (CHMP) protein family. The protein is part of the endosomal sorting complex required for transport (ESCRT) complex III (ESCRT-III), which functions in the sorting of endocytosed cell-surface receptors into multivesicular endosomes. The ESCRT machinery also functions in the final abscisson stage of cytokinesis and in the budding of enveloped viruses such as HIV-1. The three proteins of the CHMP4 subfamily interact with programmed cell death 6 interacting protein (PDCD6IP, also known as ALIX), which also functions in the ESCRT pathway. The CHMP4 proteins assemble into membrane-attached 5-nm filaments that form circular scaffolds and promote or stabilize outward budding. These polymers are proposed to help generate the luminal vesicles of multivesicular bodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP4B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHMP4B
Gene Accession No. Q9H444
Gene Alias 2010012F05Rik; C20orf178; C76846; charged multivesicular body protein 4b; CHMP4A; CHMP4B; chromatin modifying protein 4B; chromatin-modifying protein 4b; CTPP3; CTRCT31; dJ553F4.4; hSnf7-2; hVps32-2; RGD1309846; RGD1565889; Shax1; SNF7; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; Snf7-2; Vacuolar protein sorting-associated protein 32-2; vacuolar protein-sorting-associated protein 32-2; Vps32-2; VPS32B
Gene Symbols CHMP4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHMP4B. Recombinant protein control fragment (Product #RP-99355).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128866
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.