Learn More
Invitrogen™ CHMP4B Polyclonal Antibody

Description
Positive test controls include: HeLa, U-251MG, Mouse spleen, Rat heart. The target is usually found in the following locations: Cytoplasm, Late endosome membrane, Peripheral membrane protein, cytosol. Immunogen sequence: MSVFGKLFGA GGGKAGKGGP TPQEAIQRLR DTEEMLSKKQ EFLEKKIEQE LTAAKKHGTK NKRAALQALK RKKRYEKQLA QIDGTLSTIE FQREALENAN TNTEVLKNMG YAAKAMKAAH DNMDIDKVDE LMQDIADQQE LAEEISTAIS KPVGFGEEFD EDELMAELEE LEQEELDKNL LEISGPETVP LPNVPSIALP SKPAKKKEEE DDDMKELENW AGSM.
Specifications
Specifications
| Antigen | CHMP4B |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.59 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | CHMP4B |
| Gene Accession No. | Q9D8B3, Q9H444 |
| Gene Alias | 2010012F05Rik; C20orf178; C76846; charged multivesicular body protein 4b; CHMP4A; CHMP4B; chromatin modifying protein 4B; chromatin-modifying protein 4b; CTPP3; CTRCT31; dJ553F4.4; hSnf7-2; hVps32-2; RGD1309846; RGD1565889; Shax1; SNF7; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; Snf7-2; Vacuolar protein sorting-associated protein 32-2; vacuolar protein-sorting-associated protein 32-2; Vps32-2; VPS32B |
| Gene Symbols | CHMP4B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.