Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP4C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554889 Supplier Invitrogen™ Supplier No. PA554889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSKLGKFFKG GGSSKSRAAP SPQEALVRLR ETEEMLGKKQ EYLENRIQRE IALAKKHGTQ N Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

CHMP4C belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP4C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHMP4C
Gene Accession No. Q96CF2
Gene Alias 2010012P02Rik; 2210015K02Rik; 2310010I16Rik; Charged multivesicular body protein 4c; Chmp4c; chromatin modifying protein 4C; chromatin-modifying protein 4c; hSnf7-3; hVps32-3; Shax3; SNF7 homolog associated with Alix 3; Snf7 homologue associated with Alix 3; SNF7-3; Vacuolar protein sorting-associated protein 32-3; vps32-3; VPS32C
Gene Symbols CHMP4C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHMP4C. Recombinant protein control fragment (Product #RP-93864).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92421
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.