Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP5 Monoclonal Antibody (OTI5C7)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527009

Catalog No. PIMA527009


Only null left
Add to Cart

Description

Description

CHMP5 Monoclonal Antibody for Western Blot, IHC (P)

CHMP5 belongs to the chromatin-modifying protein/charged multivesicular body protein family. These proteins are components of ESCRT-III, a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies. Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHMP5
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NZZ3
CHMP5
Full length human recombinant protein of CHMP5 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI5C7
Unconjugated
CHMP5
2210412K09Rik; apoptosis-related protein PNAS-2; AW545668; C9orf83; CGI-34; Charged multivesicular body protein 5; CHMP5; chromatin modifying protein 5; chromatin-modifying protein 5; HGNC:26942; HSPC177; hVps60; likely ortholog of H. sapiens SNF7 domain containing 2; PNAS-114; PNAS-2; SNF7 domain containing 2; SNF7 domain-containing protein 2; SNF7DC2; Vacuolar protein sorting-associated protein 60; Vps60
Mouse
Affinity Chromatography
RUO
51510
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.