Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521787 Supplier Invitrogen™ Supplier No. PA521787
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse stomach. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (92%), Pig (100%), Chicken (94%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CHMP5 belongs to the chromatin-modifying protein/charged multivesicular body protein family. These proteins are components of ESCRT-III, a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies. Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP5
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene CHMP5
Gene Accession No. Q9D7S9, Q9NZZ3
Gene Alias 2210412K09Rik; apoptosis-related protein PNAS-2; AW545668; C9orf83; CGI-34; Charged multivesicular body protein 5; CHMP5; chromatin modifying protein 5; chromatin-modifying protein 5; HGNC:26942; HSPC177; hVps60; likely ortholog of H. sapiens SNF7 domain containing 2; PNAS-114; PNAS-2; SNF7 domain containing 2; SNF7 domain-containing protein 2; SNF7DC2; Vacuolar protein sorting-associated protein 60; Vps60
Gene Symbols CHMP5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CHMP5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51510, 76959
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.