Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215889
Description
CHMP6 Polyclonal specifically detects CHMP6 in Human samples. It is validated for Western Blot.Specifications
CHMP6 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
charged multivesicular body protein 6, chromatin modifying protein 6, Chromatin-modifying protein 6, FLJ11749, Vacuolar protein sorting-associated protein 20, Vps20, VPS20hVps20 | |
Rabbit | |
23 kDa | |
0.1 mL | |
Signal Transduction | |
79643 | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q96FZ7 | |
CHMP6 | |
Recombinant protein encompassing a sequence within the center region of human CHMP6. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (95%), Chicken (81%), Rat (96%), Bovine (90%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction