Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555084 Supplier Invitrogen™ Supplier No. PA555084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFGNECLNKM HQVMSIEEVE RILDETQEAV EYQRQIDELL AGSFTQEDED AILEELSAIT QEQIELPEVP SEPLPEKIPE NVPVK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.

CHMP6 belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHMP6
Gene Accession No. Q96FZ7
Gene Alias 2400004G01Rik; Charged multivesicular body protein 6; Chmp6; chromatin modifying protein 6; chromatin-modifying protein 6; hVps20; novel SNF7 protein; RGD1565325; vacuolar protein sorting-associated protein 20; VPS20
Gene Symbols CHMP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHMP6. Recombinant protein control fragment (Product #RP-93438).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79643
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.